Potassium-2,5-dihydroxybenzenesulphonate, Sigma-Aldrich®
Catalog # SIALH18402-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:228.27 g/mol
:M-Clarity™ quality level = MQ100
EINECS:244-584-7
CAS Number:21799-87-1
Formula:C₆H₅KO₅S
MDL Number:MFCD00007475