Order Entry
Export
ContactUsLinkComponent
2,4-Diaminophenol dihydrochloride, Sigma-Aldrich®
2,4-Diaminophenol dihydrochloride, Sigma-Aldrich®
Catalog # SIAL230103-100G
Supplier:  Merck
CAS Number:  
2,4-Diaminophenol dihydrochloride, Sigma-Aldrich®
Catalog # SIAL230103-100G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :197.06 g/mol
ADR:6.1,III
 :M-Clarity™ quality level = MQ100
EINECS:205-279-4
UN:2811
CAS Number:137-09-7
Formula:C₆H₁₀Cl₂N₂O
MDL Number:MFCD00012979