

2,4-Diaminophenol dihydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:137-09-7
:
Formula:C₆H₁₀Cl₂N₂O
MDL Number:MFCD00012979
:197.06 g/mol
Melting Pt:222 °C (dec.)
Storage Temperature:Ambient
EINECS:205-279-4
UN:2811
ADR:6.1,III
:GHS08
:GHS05
Specification Test Results
- Appearance:Beige to grey-green Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:≥97.5 %