Order Entry
Export
Orders LinkContactUsLinkComponent
Triclosan

Triclosan

Glass bottle for solids, 100 g
Catalog # MOLEM35047938
Supplier:   Molekula
Triclosan
Catalog # MOLEM35047938
Supplier:  Molekula
CAS Number:  3380-34-5
Triclosan

Glass bottle for solids, 100 g

CAS Number:  3380-34-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol
  • CAS Number:3380-34-5
  • Formula:C₁₂H₇Cl₃O₂
  • MDL Number:MFCD00800992
  • Molecular Weight:289.54 g/mol
  • EINECS:222-182-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications