Order Entry
Export
Orders LinkContactUsLinkComponent
 

Triclosan ≥99%

Triclosan ≥99%
Certificates
Triclosan ≥99%
Triclosan ≥99%

3 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 3380-34-5 Molecular Formula: C12H7Cl3O2 Formula Weight: 289.55 Melting Point: 56-59° Beilstein Reference: 2057142 Merck Reference: 14,9657 UN No.: 3077 MDL No.: MFCD00800992

Synonyms:  5-Chloro-2-(2,4-dichlorophenoxy)phenol, 5-Chloro-2-(2, 4-dichlorophenoxy) phenol
  • CAS Number:3380-34-5
  • Formula:C₁₂H₇Cl₃O₂
  • MDL Number:MFCD00800992
  • Molecular Weight:289.54 g/mol
  • Merck Index:14,9657
  • Melting Pt:56…59 °C
  • Storage Temperature:Ambient
  • EINECS:222-182-2
  • ADR:9,III
Product Family Options

Product Information