Tributyl acetylcitrate
200 mg
Catalog # 1009901.
Supplier: USP
Jump to:
Product Details & Documents
Synonyms: Tributyl O-Acetylcitrate, Acetyltributylcitrate, Acetyl tributyl citrate, Tributyl 2-acetoxypropane-1,2,3-tricarboxylate, O-Acetylcitric acid tributyl ester
- CAS Number:77-90-7
- Formula:C₂₀H₃₄O₈
- MDL Number:MFCD00043554
- Molecular Weight:402.49 g/mol
- EINECS:201-067-0
Specifications
Specifications
- Pk:200 mg



