D-Panthenol 98+%
:81-13-0
:
:C₉H₁₉NO₄
:MFCD00065006
:205.25 g/mol
:118…120 °C (2.7Pa)
:Refrigerator
:201-327-3
- Appearance:Clear colorless to light yellow Viscous liquid or solid
- Infrared spectrum:Conforms
- Refractive index:1.4950 to 1.5020 (20°C, 589 nm)
- Assay:≥98.0 %
- Water:≤1.0 % (K.F.)
- Specific optical rotation:+29.0° to +32.0° (20°C, 589 nm) (c=5, H2O)