Order Entry
Export
ContactUsLinkComponent
2-(3-Chloropropyl)-1,3-dioxolane, Sigma-Aldrich®
2-(3-Chloropropyl)-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIAL26235-25ML-F
Supplier:  Merck
CAS Number:  
2-(3-Chloropropyl)-1,3-dioxolane, Sigma-Aldrich®
Catalog # SIAL26235-25ML-F
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

About this item

 :
 :150.6 g/mol
CAS Number:16686-11-6
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00043135