Order Entry
Export
ContactUsLinkComponent
2-(3-Chloropropyl)-1,3-dioxolane
2-(3-Chloropropyl)-1,3-dioxolane
Catalog # MOLE33355557-5G
Supplier:  Molekula
CAS Number:  
2-(3-Chloropropyl)-1,3-dioxolane
Catalog # MOLE33355557-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :150.6 g/mol
CAS Number:16686-11-6
Formula:C₆H₁₁ClO₂
MDL Number:MFCD00043135