2-(2-Bromoethyl)-1,3-dioxolane
Catalog # MOLEM85647981
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:181.03 g/mol
EINECS:242-551-1
CAS Number:18742-02-4
Formula:C₅H₉BrO₂
MDL Number:MFCD00003216