Order Entry
Export
Orders LinkContactUsLinkComponent
 

Melatonin, Sigma-Aldrich®

Supplier:   Merck
Melatonin, Sigma-Aldrich®
Supplier:  Merck
Certificates
Melatonin, Sigma-Aldrich®

4 Options Available Below

Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:200-797-7
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 71,90 €
    71,90 €
    1 * 250 mg
       71,90 €

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 135,00 €
    135,00 €
    1 * 1 g
       135,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 551,00 €
    551,00 €
    1 * 5 g
       551,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 942,00 €
    942,00 €
    1 * 10 g
       942,00 €