Melatonin
Glass bottle for solids, 1 g
Catalog # MOLE88168715-1G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
- CAS Number:73-31-4
- Formula:C₁₃H₁₆N₂O₂
- MDL Number:MFCD00005655
- Molecular Weight:232.28 g/mol
- EINECS:200-797-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



