
2,5-Dihydroxy-p-benzoquinone ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:615-94-1
:
Formula:C₆H₄O₄
MDL Number:MFCD00001598
:140.1 g/mol
Melting Pt:∼ 220 °C (subl.)
EINECS:210-454-3
:GHS07
Storage Temperature:Ambient