Order Entry
Export
ContactUsLinkComponent
2,6-Dimethoxy-p-benzoquinone 97%
Irritant
Health
2,6-Dimethoxy-p-benzoquinone 97%
Certificates
undefined
Irritant
Health
2,6-Dimethoxy-p-benzoquinone 97%

Specification Test Results

  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    253 to 257 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information