1-Amino-3,3-diethoxypropane, Sigma-Aldrich®
Catalog # SIALA8597-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:147.22 g/mol
UN:2735
ADR:8,I
CAS Number:41365-75-7
Formula:C₇H₁₇NO₂
MDL Number:MFCD00142581