2,5-Piperazinedione 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:106-57-0
:
Formula:C₄H₆N₂O₂
MDL Number:MFCD00006009
:114.1 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:203-411-5
Specification Test Results
- Appearance:White to light yellow to beige Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥97.5 % (ex N)
- Loss on drying:≤0.5 % (105°C, 4 h)