Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L(+)-Ornithine hydrochloride
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
L(+)-Ornithine hydrochloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
3184-13-2
:
Formula:
C₅H₁₂N₂O₂·HCl
MDL Number:
MFCD00064562
:
168.62 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
221-678-6