2,6-Dichloro-p-benzoquinone, Sigma-Aldrich®
Catalog # SIAL431982-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:176.99 g/mol
:M-Clarity™ quality level = MQ200
EINECS:211-810-0
CAS Number:697-91-6
Formula:C₆H₂Cl₂O₂
MDL Number:MFCD00037159