

2,6-Dichloro-p-benzoquinone ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:697-91-6
:
Formula:C₆H₂Cl₂O₂
MDL Number:MFCD00037159
:176.99 g/mol
Melting Pt:119…121 °C
EINECS:211-810-0
:GHS06
:GHS07
Storage Temperature:Ambient