2,5-Dichloro-p-benzoquinone, Sigma-Aldrich®
Catalog # SIAL431974-10G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
:176.99 g/mol
EINECS:210-453-8
CAS Number:615-93-0
Formula:C₆H₂Cl₂O₂
MDL Number:MFCD00041736