2,5-Dibromohydroquinone, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 2,5-Dibromobenzene-1,4-diol
- CAS Number:14753-51-6
- Formula:C₆H₄Br₂O₂
- MDL Number:MFCD00192664
- Molecular Weight:267.9 g/mol
2 Options Available Below
Product Details & Documents
Synonyms: 2,5-Dibromobenzene-1,4-diol
- CAS Number:14753-51-6
- Formula:C₆H₄Br₂O₂
- MDL Number:MFCD00192664
- Molecular Weight:267.9 g/mol



