




Hydroxylamine solution 50% (w/w) in water, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7803-49-8
:
Formula:H₃NO
MDL Number:MFCD00044522
:33.03 g/mol
ADR:8,III
:M-Clarity™ quality level = MQ200
EINECS:232-259-2
UN:3266
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient