2,4,6-Trichlorophenol
Glass bottle for solids, 100 mg
Catalog # ACCUA026
Supplier: AccuStandard
Jump to:
Product Details & Documents
- CAS Number:88-06-2
- Formula:C₆H₃Cl₃O
- MDL Number:MFCD00002172
- Molecular Weight:197.45 g/mol
- Merck Index:12,09773
- Boiling Pt:244.5 °C (1013 hPa)
- Melting Pt:∼66 °C
- Density:1.675 g/cm³ (25 °C)
- Flash Pt:99 °C
- Storage Temperature:Ambient
- EINECS:201-795-9
- UN:2020
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



