Order Entry
Export
Orders LinkContactUsLinkComponent
 

4-Chlorophenol

Glass bottle for solids, 100 mg
Catalog # ACCUA015
Irritant
Environment
Supplier:   AccuStandard
4-Chlorophenol
Catalog # ACCUA015
Supplier:  AccuStandard
CAS Number:  106-48-9
4-Chlorophenol

Glass bottle for solids, 100 mg

CAS Number:  106-48-9
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Synonyms:  p-Chlorophenol
  • CAS Number:106-48-9
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002318
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:220 °C (1013 hPa)
  • Melting Pt:41…44 °C
  • Density:1.31 g/cm³ (20 °C)
  • Flash Pt:121 °C
  • Storage Temperature:Ambient
  • EINECS:203-402-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg

Specifications