Order Entry
Export
ContactUsLinkComponent
3-Chlorophenol
Irritant
Environment
3-Chlorophenol
Catalog # ACCUA014
Supplier:  AccuStandard
CAS Number:  
Irritant
Environment
3-Chlorophenol
Catalog # ACCUA014
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
Merck Index:13,02173
Boiling Pt:∼214 °C (1013 hPa)
Melting Pt:31…34 °C
Density:1.245 g/cm³ (20 °C)
Flash Pt:97 °C
Storage Temperature:Refrigerator
EINECS:203-582-6
UN:2020
ADR:6.1,III
 :128.56 g/mol
 :GHS07
 :GHS09
CAS Number:108-43-0
Formula:C₆H₅ClO
MDL Number:MFCD00002256