

1,1-Dimethoxyacetone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:6342-56-9
:
Formula:C₅H₁₀O₃
MDL Number:MFCD00008758
:118.13 g/mol
Boiling Pt:143 °C (1013 hPa)
Melting Pt:–57 °C
Density:1.0041 g/cm³ (20 °C)
EINECS:228-735-4
ADR:3,III
:M-Clarity™ quality level = MQ200
UN:1224
:GHS02
:GHS07