Storage containers, Tubby®
Catalog # HEATHS23453
Supplier: HEATHROW
Specifications
- W×D×H:330×200×115 mm
- Pk:5
Specifications
About this item
Constructed of high-impact polypropylene, these sturdy storage tubs with lid and dividers are designed to help organize a laboratory.
- Removable dividers can create as big or as small a compartment as needed
- The lid snaps in place to secure the contents and for safe transport
- Perfect for keeping tubes, tips, pipettes, gloves, and other small items organized and out of the way