ContactUsLinkComponent
Accessories for SCAT Politainer
Accessories for SCAT Politainer
  SCTV107338
 :  S.C.A.T.
undefined
Accessories for SCAT Politainer
  SCTV107338
 :  S.C.A.T.
  • For:
    Foldable politainer with GL 38
  • Description:
    Thread adapter, PTFE, GL 38 on GL 45
  • Seq:
    04
  • Pk:
    1
The SCAT Politainer - also known as folding canisters - is a combination of a flexible plastic bag and a stable UN-approved outer carton. Storage in the unfilled state is possible in a very small space and after filling, safe storage or transport is guaranteed.
  • Thread to adjust the thread from GL38 to GL45
  • Especially suitable for SCAT Politainer
SCAT Politainers are an ideal way of not having to dispense with safe storage even in limited spaces.