Goggles, Honeywell LG
The impact resistant polycarbonate lens offers an excellent protection against flying particles, coarse dusts, liquid droplets, mists and sprays.
- Lightweight design and large field of vision
- Soft seal and easily adjustable headband: Fits a wide variety of workers
- With or without ventilation
The acetate, anti-fog lens and neoprene headband offers superior chemical resistance allowing the use in a broad range of chemical industries.
Weight: 75 to 79 g
: EN 166