Order Entry
Export
ContactUsLinkComponent
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Catalog # MOLEM22590378
Supplier:  Molekula
CAS Number:  
Benzo-15-crown-5 (benzo-1,4,7,10,13-pentaoxacyclopentadecane)
Catalog # MOLEM22590378
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :268.31 g/mol
EINECS:237-947-6
CAS Number:14098-44-3
Formula:C₁₄H₂₀O₅
MDL Number:MFCD00005945