Order Entry
Export
ContactUsLinkComponent
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane), Sigma-Aldrich®
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane), Sigma-Aldrich®
Catalog # SIAL11378-1G
Supplier:  Merck
CAS Number:  
1-Aza-12-crown-4 (1,4,7-trioxa-10-azacyclododecane), Sigma-Aldrich®
Catalog # SIAL11378-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.23 g/mol
EINECS:255-549-0
CAS Number:41775-76-2
Formula:C₈H₁₇NO₃
MDL Number:MFCD00134269