

Acridine 97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:260-94-6
:
Formula:C₁₃H₉N
MDL Number:MFCD00005025
:179.22 g/mol
Boiling Pt:346 °C (lit.)
Melting Pt:107...110 °C (lit.)
EINECS:205-971-6
UN:2713
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS07
:GHS09
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Light Yellow to Very Dark Yellow and Light Beige to Beige and Light Brown to Brown
- Appearance (Form):Powder or Crystals
- Infrared spectrum:Conforms to Structure
- Purity (Titration by HCL04):96.5 - 103.5%
- Purity (HPLC):≥96.5%