o-Phenylenediacetonitrile 99+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,2-Phenylenediacetonitrile, 1,2-Bis(cyanomethyl)-benzene
CAS Number:613-73-0
:
Formula:C₁₀H₈N₂
MDL Number:MFCD00001905
:156.19 g/mol
Melting Pt:58…61 °C
Storage Temperature:Ambient
EINECS:210-351-3
UN:3439
ADR:6.1,III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to yellow to beige Crystalline powder or crystals (May contain yellow to orange chunks)
- Infrared spectrum:Conforms
- Melting point:57 to 61 °C
- GC:≥99.0 %
- Residue after evaporation:≤0.001 %
- Filtered to 0.2 micrometer:Filtered