Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,2-Bis(hydroxymethyl)propionic acid
2,2-Bis(hydroxymethyl)propionic acid, Sigma-Aldrich®
2,2-Bis(hydroxymethyl)propionic acid, Sigma-Aldrich®
2,2-Bis(hydroxymethyl)propionic acid, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
2,2-Bis(hydroxymethyl)propionic acid, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
4767-03-7
:
Formula:
C₅H₁₀O₄
MDL Number:
MFCD00004199
:
134.13 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
225-306-3