
2,2-Bis(hydroxymethyl)propionic acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4767-03-7
:
Formula:C₅H₁₀O₄
MDL Number:MFCD00004199
:134.13 g/mol
Melting Pt:185…190 °C
Flash Pt:>150 °C
EINECS:225-306-3
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Titration with NaOH:≥97.5 %