(±)-Camphorquinone, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
CAS Number:10373-78-1
:
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00064160
:166.22 g/mol
:M-Clarity™ quality level = MQ100
EINECS:233-814-1
Specifications
- Environmentally Preferable: