(±)-Camphorquinone
Catalog # MOLEM60570677
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
:
:166.22 g/mol
EINECS:233-814-1
CAS Number:10373-78-1
Formula:C₁₀H₁₄O₂
MDL Number:MFCD00064160