Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sesamol (3,4-(Methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
Supplier:
Roth Carl
Certificates
Expand image
Sesamol (3,4-(Methylenedioxy)phenol)
Supplier:
Roth Carl
About this item
CAS Number:
533-31-3
:
Formula:
C₇H₆O₃
MDL Number:
MFCD00005827
:
138.12 g/mol
EINECS:
208-561-5