Order Entry
Export
ContactUsLinkComponent
Sesamol (3,4-(Methylenedioxy)phenol)
Sesamol (3,4-(Methylenedioxy)phenol)
Catalog # MOLE10825829-25G
Supplier:  Molekula
CAS Number:  
Sesamol (3,4-(Methylenedioxy)phenol)
Catalog # MOLE10825829-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :138.12 g/mol
EINECS:208-561-5
CAS Number:533-31-3
Formula:C₇H₆O₃
MDL Number:MFCD00005827