Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-(Methylenedioxy)aniline
3,4-(Methylenedioxy)aniline, Sigma-Aldrich®
3,4-(Methylenedioxy)aniline, Sigma-Aldrich®
3,4-(Methylenedioxy)aniline, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
3,4-(Methylenedioxy)aniline, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
14268-66-7
:
Formula:
C₇H₇NO₂
MDL Number:
MFCD00005832
:
137.14 g/mol
EINECS:
238-161-6
UN:
2811
ADR:
6.1,III