3,4-(Methylenedioxy)aniline
Catalog # MOLEM49406147
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:137.14 g/mol
EINECS:238-161-6
UN:2811
ADR:6.1,III
CAS Number:14268-66-7
Formula:C₇H₇NO₂
MDL Number:MFCD00005832