Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(±)-Ethyl 2,3-dibromopropionate
(±)-Ethyl 2,3-dibromopropionate
(±)-Ethyl 2,3-dibromopropionate
MOLE34893089-100G
:
Molekula
:
(±)-Ethyl 2,3-dibromopropionate
MOLE34893089-100G
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
259.93 g/mol
:
222-941-8
:
3674-13-3
:
C₅H₈Br₂O₂
:
MFCD00000212