2,2-Dichloropropionic acid
Catalog # EHERC11949500
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:142.97 g/mol
EINECS:200-923-0
CAS Number:75-99-0
Formula:C₃H₄Cl₂O₂
MDL Number:MFCD00004178