0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
1,4-Diethylpiperazine
1,4-Diethylpiperazine ≥98%
1,4-Diethylpiperazine ≥98%
:
Thermo Fisher Scientific
1,4-Diethylpiperazine ≥98%
:
Thermo Fisher Scientific
:
6483-50-7
:
:
C₈H₁₈N₂
:
MFCD00126900
:
142.24 g/mol
:
229-341-5
:
GHS07
:
Ambient
msds_ALFAB24743
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years