Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 3,3-diethoxypropionate
Ethyl-3,3-diethoxypropionate, Sigma-Aldrich®
Ethyl-3,3-diethoxypropionate, Sigma-Aldrich®
Ethyl-3,3-diethoxypropionate, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Ethyl-3,3-diethoxypropionate, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
10601-80-6
:
Formula:
C₉H₁₈O₄
MDL Number:
MFCD00009865
:
190.24 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
234-223-1