Methyl anthranilate, Sigma-Aldrich®
Catalog # SIAL236454-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:151.16 g/mol
Merck Index:12,06099
Boiling Pt:237 °C (1013 hPa)
Melting Pt:24 °C
Density:1.16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-132-4
CAS Number:134-20-3
Formula:C₈H₉NO₂
MDL Number:MFCD00007710