2-Adamantanol 98%
ACRO167310050
:
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:258 to 265 °C (Subl)
- GC:≥97.5 %
:
:152.24 g/mol
:258…265 °C
:1,07 g/cm³ (25 °C)
:Ambient
:211-846-7
:700-57-2
:C₁₀H₁₆O
:MFCD00074744