

2,3-Dimethylthiophene 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:632-16-6
:
Formula:C₆H₈S
MDL Number:MFCD00130081
:112.2 g/mol
Boiling Pt:141.6 °C (1013 hPa)
Melting Pt:–49 °C
Density:1,0021 g/cm³ (20 °C)
Flash Pt:>1 °C
Storage Temperature:Ambient
EINECS:211-170-2
UN:1993
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5170 to 1.5210 (20°C, 589 nm)
- GC:≥96.0 %