Order Entry
Export
Orders LinkContactUsLinkComponent
2-Hexylthiophene ≥98%

2-Hexylthiophene ≥98%

2-Hexylthiophene ≥98%
Certificates
2-Hexylthiophene ≥98%
2-Hexylthiophene ≥98%

2 Options Available Below

 

Purity: 98% CAS No.: 18794-77-9 Molecular Formula: C10H16S Formula Weight: 168.30 Flash Point: 100°(212°F) Refractive Index: 1.4960 Boiling Point: 228-230° Beilstein Reference: 110220 MDL No.: MFCD00022535

  • CAS Number:18794-77-9
  • Formula:C₁₀H₁₆S
  • MDL Number:MFCD00022535
  • Molecular Weight:168.3 g/mol
  • Boiling Pt:228…230 °C
  • Density:0.935
  • Flash Pt:100 °C (212 °F)
  • Storage Temperature:Ambient
  • EINECS:242-579-4
Product Family Options

Product Information