2,5-Dichlorothiophene
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:3172-52-9
:
Formula:C₄H₂Cl₂S
MDL Number:MFCD00005423
:153.03 g/mol
Boiling Pt:162 °C (1013 hPa)
Melting Pt:–40 °C
Density:1.4473 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:221-638-8
UN:2929
ADR:6.1,II